Få mere at vide
Hydroxyphenamate, Mikromol™
Beskrivelse
Hydroxyphenamate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Hydroxyphenamate |
| CAS | 50-19-1 |
| Analyt- eller komponentnavne | Hydroxyphenamate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C11 H15 N O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 1,2-Butanediol, 2-phenyl-, 1-carbamate, 2-Hydroxy-2-phenylbutyl carbamate, AL 0361, Hydroxyphenamate, Listica, NSC 108034, Oxyfenamate, Oxyphenamate, P 301, Carbamic acid, beta-ethyl-beta-hydroxyphenethyl ester (7CI), beta-Ethyl-beta-hydroxyphenethyl carbamate |
| InChI formel | InChI=1S/C11H15NO3/c1-2-11(14,8-15-10(12)13)9-6-4-3-5-7-9/h3-7,14H,2,8H2,1H3,(H2,12,13) |
| SMIL | CCC(O)(COC(=O)N)c1ccccc1 |
| IUPAC navn | (2-hydroxy-2-phenylbutyl) carbamate |
| Vis mere |