missing translation for 'onlineSavingsMsg'
Få mere at vide

Hydroxyphenamate, LoGiCal

Artikelnummer. 30459690
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459690 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30459690 Leverandør LoGiCal Leverandørnr. LGCFOR3109.00

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Especificaciones

Kemisk navn eller materiale Hydroxyphenamate
CAS 50-19-1
Analyt- eller komponentnavne Hydroxyphenamate
Forsendelsestilstand Room Temperature
Molekylær formel C11 H15 N O3
Certificeringer/compliance ISO 9001
Synonym 1,2-Butanediol, 2-phenyl-, 1-carbamate, 2-Hydroxy-2-phenylbutyl carbamate, AL 0361, Hydroxyphenamate, Listica, NSC 108034, Oxyfenamate, Oxyphenamate, P 301, Carbamic acid, beta-ethyl-beta-hydroxyphenethyl ester (7CI), beta-Ethyl-beta-hydroxyphenethyl carbamate
InChI formel InChI=1S/C11H15NO3/c1-2-11(14,8-15-10(12)13)9-6-4-3-5-7-9/h3-7,14H,2,8H2,1H3,(H2,12,13)
Løsningstype Neat
SMIL CCC(O)(COC(=O)N)c1ccccc1
IUPAC navn (2-hydroxy-2-phenylbutyl) carbamate
Industry Type Clinical Testing
Molekylvægt (g/mol) 209.24
Mængde 10 mg
Formel vægt 209.1052 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.