Få mere at vide
Hydroxymethyl Tolperisone-d10 Hydrochloride, TRC
Artikelnummer. 30406363
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30406363 50 mg
30405851 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406363 Leverandør Toronto Research Chemicals Leverandørnr. TRCH82552750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346605-29-5
Molekylær formel C16H14D10ClNO2
Molekylvægt (g/mol) 307.88
Synonym 1-[4-(Hydroxymethyl)phenyl]-2-methyl-3-(1-piperidinyl-d10)-1-propanone Hydrochloride
IUPAC navn 3-(2,2,3,3,4,4,5,5,6,6-decadeuteriopiperidin-1-yl)-1-[4-(hydroxymethyl)phenyl]-2-methylpropan-1-one;hydrochloride
SMIL Cl.[2H]C1([2H])N(CC(C)C(=O)c2ccc(CO)cc2)C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
Tekniske data
Kemisk navn eller materiale Hydroxymethyl Tolperisone-d10 Hydrochloride
Mængde 50 mg
InChI formel InChI=1S/C16H23NO2.ClH/c1-13(11-17-9-3-2-4-10-17)16(19)15-7-5-14(12-18)6-8-15;/h5-8,13,18H,2-4,9-12H2,1H3;1H/i2D2,3D2,4D2,9D2,10D2;
Formel vægt 307.2123