Få mere at vide
Hydroxylaminsulfat, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Hydroxylamine Sulfate |
| CAS | 10039-54-0 |
| Molekylær formel | H8N2O6S |
| Mængde | 10 g |
| Synonym | Bis(hydroxylamine) Sulfate, Bis(hydroxylammonium) Sulfate, Di(hydroxylamine) Sulfate, Envision MLB 2030, Hydroxyl Ammonium Sulfate ((HONH3)2SO4), Hydroxylamine Neutral Sulfate, Hydroxylammonium Sulfate, Lanasane LAB, Oxammonium Sulfate |
| InChI formel | InChI=1S/2H4NO.H2O4S/c2*1-2;1-5(2,3)4/h2*2H,1H3;(H2,1,2,3,4)/q2*+1;/p-2 |
| SMIL | NO.NO.OS(=O)(=O)O |
| IUPAC navn | hydroxylamin; Svovlsyre |
| Molekylvægt (g/mol) | 164.14 |
| Formel vægt | 164.0103 |