missing translation for 'onlineSavingsMsg'
Få mere at vide

Hydroxylamine, 50% aq. soln., Thermo Scientific™

335.67 DKK

Kemiske identifikatorer

CAS 7803-49-8
Molekylær formel H3NO
Molekylvægt (g/mol) 33.03
MDL nummer MFCD00044522
InChI nøgle AVXURJPOCDRRFD-UHFFFAOYSA-N
Synonym oxammonium, nitroxide, hydroxyamine, nh2oh, oxyammonia, nitrinous acid, unii-2fp81o2l9z, hsdb 579, dihydridohydroxidonitrogen, arninoalcohol
PubChem CID 787
ChEBI CHEBI:15429
IUPAC navn hydroxylamine
SMIL NO
Se flere specifikationer

Produkter 1
Produktkode Brand Mængde Pris Antal & tilgængelighed  
Produktkode Brand Mængde Pris Antal & tilgængelighed  
15453237
Dokumentvisning
Thermo Scientific Alfa Aesar
B22202.AE
100mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ikke relevant Se alternative produkter
Spar op til 
»
Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Hydroxylamine is a reactant used for the preparation of hydroxyaminoguanidines which are anti-cancer agents, carboxamide derivatives of ofloxacin and human immunodeficiency virus (HIV) integrase inhibitors. It reacts with chlorosulfonic acid to give hydroxylamine-O-sulfonic acid, a useful reagent in the synthesis of caprolactam. It is commonly used as a reducing agent in myriad of organic and inorganic reactions. It acts as an antioxidant for fatty acids. It is widely used to prepare oximes, an important functional group.

Solubility
Miscible with water, liquid ammonia and methanol. Slightly miscible with ether, benzene, carbon disulfide and chloroform.

Notes
It can violently decompose at elevated temperatures. May be shock-sensitive if dry. Do not heat over 500C. Incompatible with bases, oxidizing agents, nitrates, nitrites, heavy metals, rubber, metals, epoxides and polyester.
Tekniske data

Kemiske identifikatorer

7803-49-8
33.03
AVXURJPOCDRRFD-UHFFFAOYSA-N
787
hydroxylamine
H3NO
MFCD00044522
oxammonium, nitroxide, hydroxyamine, nh2oh, oxyammonia, nitrinous acid, unii-2fp81o2l9z, hsdb 579, dihydridohydroxidonitrogen, arninoalcohol
CHEBI:15429
NO

Tekniske data

Hydroxylamine
7803-49-8
H3NO
MFCD00044522
1098214
14,4828
Miscible with water,liquid ammonia and methanol. Slightly miscible with ether,benzene,carbon disulfide and chloroform.
NO
33.03
CHEBI:15429
1.394
aq. soln.
0.5
NH2OH
UN3266
None
oxammonium, nitroxide, hydroxyamine, nh2oh, oxyammonia, nitrinous acid, unii-2fp81o2l9z, hsdb 579, dihydridohydroxidonitrogen, arninoalcohol
AVXURJPOCDRRFD-UHFFFAOYSA-N
hydroxylamine
787
33.03
1.120
Videoer
Sikkerhed og håndtering

Sikkerhed og håndtering

GHS H Statement
H318-H351-H373-H302-H317-H335-H315

P201-P202-P234-P260-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P333+P313-P362-P372-P373-P380-P390-P501c

H290-H302+H312-H315-H317-H318-H335-H351-H373

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 232-259-2

missing translation for 'rtecsNumber' : NC2975000

missing translation for 'tsca' : Yes

Anbefalet opbevaring : Ambient temperatures

SDS
Dokumenter

Dokumenter

Produktcertifikater

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.