missing translation for 'onlineSavingsMsg'
Få mere at vide

α-Hydroxyetizolam, TRC

Artikelnummer. 30350323
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30350323 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350323 Supplier Toronto Research Chemicals Supplier No. TRCC3801052.5MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale alpha-Hydroxyetizolam
CAS 64546-10-7
Molekylær formel C17 H15 Cl N4 O S
Mængde 2.5 mg
Synonym , (2-Chlorophenyl)-α,9-dimethyl-6H-Thieno[3,2-f][1,2,4]triazolo [4,3-a][1,4]diazepine-2-methanol, 4-(2-Chlorophenyl)-α,9-dimethyl-6H-Thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-2-methanol
InChI formel InChI=1S/C17H15ClN4OS/c1-9(23)14-7-12-16(11-5-3-4-6-13(11)18)19-8-15-21-20-10(2)22(15)17(12)24-14/h3-7,9,23H,8H2,1-2H3
SMIL CC(O)c1cc2C(=NCc3nnc(C)n3c2s1)c4ccccc4Cl
IUPAC navn 1-[7-(2-chlorophenyl)-13-methyl-3-thia-1,8,11,12-tetrazatricyclo[8.3.0.02,6]trideca-2(6),4,7,10,12-pentaen-4-yl]ethanol
Molekylvægt (g/mol) 358.845
Formel vægt 358.066
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.