Få mere at vide
Hydroxydiazinon, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Hydroxydiazinon |
| CAS | 29820-16-4 |
| Molekylær formel | C12 H21 N2 O4 P S |
| Mængde | 2.5 mg |
| Synonym | Phosphorothioic acid, O,O-diethyl O-[2-(1-hydroxy-1-methylethyl)-6-methyl-4-pyrimidinyl] ester, 2-Pyrimidinemethanol, 4-hydroxy-α,α,6-trimethyl-, 4-O-ester with O,O-diethyl phosphorothioate (8CI), O,O-Diethyl O-[2-(1-hydroxy-1-methylethyl)-6-methylpyrimidin-4-yl] phosphorothioate, O,O-Diethyl O-[2-(2-hydroxyisopropyl)-6-methyl-4-pyrimidinyl] phosphorothioate, O,O-Diethyl O-[2-(2'-hydroxy-2'-propyl)-4-methyl-6-pyrimidinyl]phosphorothioate |
| InChI formel | InChI=1S/C12H21N2O4PS/c1-6-16-19(20,17-7-2)18-10-8-9(3)13-11(14-10)12(4,5)15/h8,15H,6-7H2,1-5H3 |
| SMIL | CCOP(=S)(OCC)Oc1cc(C)nc(n1)C(C)(C)O |
| IUPAC navn | 2-(4-diethoxyphosphinothioyloxy-6-methylpyrimidin-2-yl)propan-2-ol |
| Molekylvægt (g/mol) | 320.345 |
| Formel vægt | 320.096 |