Få mere at vide
Hydroxychloroquine N-Oxide Dihydrochloride, Mikromol™
Beskrivelse
Hydroxychloroquine N-Oxide Dihydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Hydroxychloroquine N-Oxide Dihydrochloride |
| CAS | 2725355-06-4 |
| Analyt- eller komponentnavne | Hydroxychloroquine N-Oxide Dihydrochloride |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C18 H26 Cl N3 O2 . 2 Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl]ethyloxidoamino]ethanol dihydrochloride, 4-[(7-Chloroquinolin-4-yl)amino]-N-ethyl-N-(hydroxyethyl)pentan-1-amine N-oxide dihydrochloride, Hydroxychloroquine N-oxide dihydrochloride, Hydroxychloroquine Sulfate Impurity A as Dihydrochloride, Hydroxychloroquine Impurity A as Dihydrochloride |
| InChI formel | InChI=1S/C18H26ClN3O2.2ClH/c1-3-22(24,11-12-23)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18;;/h6-9,13-14,23H,3-5,10-12H2,1-2H3,(H,20,21);2*1H |
| SMIL | Cl.Cl.CC[N+]([O-])(CCO)CCCC(C)Nc1ccnc2cc(Cl)ccc12 |
| IUPAC navn | 4-[(7-chloroquinolin-4-yl)amino]-N-ethyl-N-(2-hydroxyethyl)pentan-1-amine oxide;dihydrochloride |
| Vis mere |