Få mere at vide
Hydroxybupropion, Lipomed
Artikelnummer. 30462702
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30462702 50 mg
1 options
Artikelnummer. 30462702 Leverandør Lipomed Leverandørnr. LPMHBP1895FB50
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Hydroxybupropion
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (±)-Hydroxybupropion
CAS 92264-81-8
Analyt- eller komponentnavne (±)-Hydroxybupropion
Forsendelsestilstand Room Temperature
Molekylær formel C13 H18 Cl N O2
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMIL CC(NC(C)(C)CO)C(=O)c1cccc(Cl)c1
IUPAC navn 1-(3-chlorophenyl)-2-[(1-hydroxy-2-methylpropan-2-yl)amino]propan-1-one
Industry Type Pharmaceutical
Molekylvægt (g/mol) 255.74
Mængde 50 mg
Formel vægt 255.103 g/mol