Få mere at vide
Hydroxybupropion (as (RS,RS)-cyclic Hemiketal), LoGiCal
Artikelnummer. 30457648
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30457648 10 mg
1 options
Artikelnummer. 30457648 Leverandør LoGiCal Leverandørnr. LGCFOR0745.14
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hydroxybupropion (as (RS,RS)-cyclic Hemiketal)
CAS 233600-52-7
Analyt- eller komponentnavne Hydroxybupropion (as (RS,RS)-cyclic Hemiketal)
Forsendelsestilstand Room Temperature
Molekylær formel C13 H18 Cl N O2
Certificeringer/compliance ISO 9001
Synonym Hydroxybupropion (as (RS,RS)-cyclic Hemiketal)
InChI formel InChI=1S/C13H18ClNO2/c1-9-13(16,17-8-12(2,3)15-9)10-5-4-6-11(14)7-10/h4-7,9,15-16H,8H2,1-3H3/t9-,13+/m0/s1
Løsningstype Neat
SMIL C[C@@H]1NC(C)(C)CO[C@@]1(O)c2cccc(Cl)c2
IUPAC navn (2S,3S)-2-(3-chlorophenyl)-3,5,5-trimethylmorpholin-2-ol
Industry Type Clinical Testing
Molekylvægt (g/mol) 255.74
Mængde 10 mg
Formel vægt 255.1026 g/mol