Få mere at vide
Hydroxyacetone-2,4-DNPH, TRC
Artikelnummer. 30137179
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137179 25 mg
30136373 250 mg
17953735 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137179 Leverandør Toronto Research Chemicals Leverandørnr. TRCH71481525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 96481-90-2
Molekylær formel C9 H10 N4 O5
Molekylvægt (g/mol) 254.2
Synonym 1-Hydroxy-2-propanone 2-(2,4-Dinitrophenyl)hydrazone;
SMIL CC(=NNc1ccc(cc1N(=O)=O)N(=O)=O)CO
Tekniske data
Kemisk navn eller materiale Hydroxyacetone-2,4-DNPH
Mængde 25 mg
InChI formel InChI=1S/C9H10N4O5/c1-6(5-14)10-11-8-3-2-7(12(15)16)4-9(8)13(17)18/h2-4,11,14H,5H2,1H3
IUPAC navn diethoxy-(4-nitrophenoxy)-sulfanylidene-λ^{5}-phosphane
Formel vægt 254.065