Få mere at vide
Hydroxy Tyrosol 4-Sulfate Sodium Salt, TRC
Artikelnummer. 30012352
Skift visning
Mængde:
0.5 mg
2.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30012352 5 mg -
30011149 0.5 mg 0.5mg
30006601 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012352 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9770155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1817821-22-9
Molekylær formel C8 H9 O6 S . Na
Molekylvægt (g/mol) 256.208
Synonym 4-(2-Hydroxyethyl)-1,2-benzenediol 1-(Hydrogen Sulfate) Sodium Salt, Hydroxy Tyrosol 4-Sulfate Sodium Salt;1,2-Benzenediol, 4-(2-hydroxyethyl)-, 1-(hydrogen sulfate), sodium salt (1:1) (ACI), 4-(2-Hydroxyethyl)-2-hydroxyphenyl sodium sulfate, [2-Hydroxy-4-(2-hydroxyethyl)phenyl]oxidanesulfonic acid sodium salt, Hydroxy tyrosol 4-sulfate sodium salt, Hydroxytyrosol 4-sulfate sodium salt, Sodium 4-(2-hydroxyethyl)-1,2-benzenediol 1-sulfate
IUPAC navn sodium;[2-hydroxy-4-(2-hydroxyethyl)phenyl] sulfate
SMIL [Na+].OCCc1ccc(OS(=O)(=O)[O-])c(O)c1
Tekniske data
Kemisk navn eller materiale Sodium 4-(2-Hydroxyethyl)-1,2-benzenediol 1-Sulfate
Mængde 5 mg
InChI formel InChI=1S/C8H10O6S.Na/c9-4-3-6-1-2-8(7(10)5-6)14-15(11,12)13;/h1-2,5,9-10H,3-4H2,(H,11,12,13);/q;+1/p-1
Formel vægt 256.002