missing translation for 'onlineSavingsMsg'
Få mere at vide

Hydroxy Torsemide, TRC

Artikelnummer. 30404075
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30404075 25 mg
30395748 10 mg
30402620 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30404075 Leverandør Toronto Research Chemicals Leverandørnr. TRCH96998025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 99300-68-2
Molekylær formel C16H20N4O4S
Molekylvægt (g/mol) 364.42
Synonym 4-[[3-(Hydroxymethyl)phenyl]amino]-N-[[(1-methylethyl)amino]carbonyl]-3-pyridinesulfonamide, AC 3534
IUPAC navn 1-[4-[3-(hydroxymethyl)anilino]pyridin-3-yl]sulfonyl-3-propan-2-ylurea
SMIL CC(C)NC(=O)NS(=O)(=O)C1=C(C=CN=C1)NC2=CC=CC(=C2)CO

Tekniske data

Kemisk navn eller materiale Hydroxy Torsemide
Mængde 25 mg
InChI formel InChI=1S/C16H20N4O4S/c1-11(2)18-16(22)20-25(23,24)15-9-17-7-6-14(15)19-13-5-3-4-12(8-13)10-21/h3-9,11,21H,10H2,1-2H3,(H,17,19)(H2,18,20,22)
Formel vægt 364.1205
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.