Få mere at vide
Hydroxy Tolbutamide-d9, TRC
Artikelnummer. 30404992
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30404992 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404992 Leverandør Toronto Research Chemicals Leverandørnr. TRCH96985210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hydroxy Tolbutamide-d9
CAS 1185112-19-9
Molekylær formel C12 2H9 H9 N2 O4 S
Mængde 10 mg
Synonym Benzenesulfonamide, N-[(butyl-1,1,2,2,3,3,4,4,4-d9-amino)carbonyl]-4-(hydroxymethyl)- (ACI), N-[(Butyl-1,1,2,2,3,3,4,4,4-d9-amino)carbonyl]-4-(hydroxymethyl)benzenesulfonamide (ACI), 4-Hydroxytolbutamide-d9, 4-Hydroxytolbutamide-D9 (N-butyl-D9), 1-(Butyl-d9)-3-[4-(hydroxymethyl)phenyl]sulfonylurea
InChI formel InChI=1S/C12H18N2O4S/c1-2-3-8-13-12(16)14-19(17,18)11-6-4-10(9-15)5-7-11/h4-7,15H,2-3,8-9H2,1H3,(H2,13,14,16)/i1D3,2D2,3D2,8D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])NC(=O)NS(=O)(=O)c1ccc(CO)cc1
IUPAC navn 1-[4-(hydroxymethyl)phenyl]sulfonyl-3-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)urea
Molekylvægt (g/mol) 295.4
Formel vægt 295.1552