Få mere at vide
Hydroxy Melphalan (>90%), TRC
Artikelnummer. 30399574
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30399574 5 mg
30395033 25 mg
30389283 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30399574 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9398505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61733-01-5
Molekylær formel C13 H19 Cl N2 O3
Molekylvægt (g/mol) 286.75
Synonym L-Phenylalanine, 4-[(2-chloroethyl)(2-hydroxyethyl)amino]-, 4-[(2-Chloroethyl)(2-hydroxyethyl)amino]-L-phenylalanine, Hydroxymelphalan, (S)-2-Amino-3-(4-((2-chloroethyl)(2-hydroxyethyl)amino)phenyl)propanoic acid, Monohydroxy Melphalan, Monohydroxymelphalan
IUPAC navn (2S)-2-amino-3-[4-[2-chloroethyl(2-hydroxyethyl)amino]phenyl]propanoic acid
SMIL N[C@@H](Cc1ccc(cc1)N(CCO)CCCl)C(=O)O
Tekniske data
Kemisk navn eller materiale hydroxy Melphalan
Mængde 5 mg
InChI formel InChI=1S/C13H19ClN2O3/c14-5-6-16(7-8-17)11-3-1-10(2-4-11)9-12(15)13(18)19/h1-4,12,17H,5-9,15H2,(H,18,19)/t12-/m0/s1
Formel vægt 286.1084