missing translation for 'onlineSavingsMsg'
Få mere at vide

Hydroxy Itraconazole, TRC

Artikelnummer. 17931915
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17931915 1 mg
17913205 10 mg
17923205 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17931915 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9436001MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 112559-91-8
Molekylær formel C35 H40 Cl2 N8 O4
Molekylvægt (g/mol) 707.65
Synonym 4-[4-[4-[4-[[2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-1-piperazinyl]phenyl]-2,4-dihydro-2-(2-hydroxy-1-methylpropyl)-3H-1,2,4-triazol-3-one, Hydroxyitraconazole, R 63373, 3H-1,2,4-Triazol-3-one, 4-[4-[4-[4-[[2-(2,4-dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-1-piperazinyl]phenyl]-2,4-dihydro-2-(2-hydroxy-1-methylpropyl)-
IUPAC navn 2-butan-2-yl-4-[4-[4-[4-[[2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-3H-1,2,4-triazol-3-ol
SMIL CCC(C)N1N=CN(C1O)c2ccc(cc2)N3CCN(CC3)c4ccc(OCC5COC(Cn6cncn6)(O5)c7ccc(Cl)cc7Cl)cc4

Tekniske data

Kemisk navn eller materiale Hydroxy Itraconazole
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C35H40Cl2N8O4/c1-3-25(2)45-34(46)44(24-40-45)29-7-5-27(6-8-29)41-14-16-42(17-15-41)28-9-11-30(12-10-28)47-19-31-20-48-35(49-31,21-43-23-38-22-39-43)32-13-4-26(36)18-33(32)37/h4-13,18,22-25,31,34,46H,3,14-17,19-21H2,1-2H3
Formel vægt 706.255
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.