Få mere at vide
Hydroxy Flutamide-d6, TRC
Artikelnummer. 30405839
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30405839 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405839 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94247725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hydroxy Flutamide-d6
CAS 223134-73-4
Molekylær formel C11H5D6F3N2O4
Mængde 25 mg
Synonym 2-Hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide-d6, Hydroxyniphtholide-d6, Sch 16423-d6, α,α,α-Trifluoro-2-methyl-4'-nitro-m- lactotoluidide-d6
InChI formel InChI=1S/C11H11F3N2O4/c1-10(2,18)9(17)15-6-3-4-8(16(19)20)7(5-6)11(12,13)14/h3-5,18H,1-2H3,(H,15,17)/i1D3,2D3
SMIL O=C(C(O)(C([2H])([2H])[2H])C([2H])([2H])[2H])NC1=CC(C(F)(F)F)=C([N+]([O-])=O)C=C1
Molekylvægt (g/mol) 298.25
Formel vægt 298.1048