Få mere at vide
Hydroxy Atrazine, TRC
Artikelnummer. 30138579
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138579 100 mg
17953505 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138579 Leverandør Toronto Research Chemicals Leverandørnr. TRCH828600100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2163-68-0
Molekylær formel C8 H15 N5 O
Molekylvægt (g/mol) 197.24
Synonym s-Triazin-2-ol, 4-(ethylamino)-6-(isopropylamino)- (7CI,8CI), 4-(Ethylamino)-6-[(1-methylethyl)amino]-1,3,5-triazin-2(1H)-one, 2-Hydroxy-4-ethylamino-6-isopropylamino-s-triazine, 2-Hydroxy-4-isopropylamino-6-ethylamino-s-triazine, 2-Hydroxyatrazine, 2-Oxy-4-(ethylamino)-6-(isopropylamino)-s-triazine, Atrazine-hydroxy, G 34048, Hydroxyatrazine, N-Ethyl-6-hydroxy-N'-(1-methylethyl)-1,3,5-triazine-2,4-diamine
IUPAC navn 4-(ethylamino)-6-(propan-2-ylamino)-1,3,5-triazin-2-ol
SMIL CCNc1nc(O)nc(NC(C)C)n1
Tekniske data
Kemisk navn eller materiale Hydroxyatrazine
Mængde 100 mg
InChI formel InChI=1S/C8H15N5O/c1-4-9-6-11-7(10-5(2)3)13-8(14)12-6/h5H,4H2,1-3H3,(H3,9,10,11,12,13,14)
Formel vægt 197.1277