Få mere at vide
Hydrocortisone Sodium Succinate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Hydrocortisone Hydrogen Succinate |
| CAS | 2203-97-6 |
| Analyt- eller komponentnavne | Hydrocortisone Hydrogen Succinate |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C25 H34 O8 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Hydrocortisone Hydrogen Succinate, 11beta,17-Dihydroxy-3,20-dioxopregn-4-en-21-yl hydrogen butanedioate, Pregn-4-ene-3,20-dione, 21-(3-carboxy-1-oxopropoxy)-11,17-dihydroxy-, (11beta)-, Cortisol, 21-(hydrogen succinate) (8CI), Cortisol, hydrogen succinate (7CI), (11beta)-21-(3-Carboxy-1-oxopropoxy)-11,17-dihydroxypregn-4-ene-3,20-dione, Cortisol 21-hemisuccinate, Cortisol hemisuccinate, Cortisol succinate, Hydrocortisone 21-(hydrogen succinate), Hydrocortisone 21-O-beta-carbonylpropionic acid, Hydrocortisone 21-hemisuccinate, Hydrocortisone 21-succinate, Hydrocortisone hemisuccinate, Hydrocortisone succinate, NSC 7576, Saxizon |
| InChI formel | InChI=1S/C25H34O8/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30/h11,16-18,22,27,32H,3-10,12-13H2,1-2H3,(H,29,30)/t16-,17-,18-,22+,23-,24-,25-/m0/s1 |
| Løsningstype | Neat |
| SMIL | C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COC(=O)CCC(=O)O)[C@@]4(C)C[C@H](O)[C@H]23 |
| Vis mere |