Få mere at vide
Hydrocodone N-Oxide, Mikromol™
Artikelnummer. 30049628
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049628 25 mg
17933226 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049628 Leverandør Mikromol Leverandørnr. MM1113.170025
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Hydrocodone N-Oxide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hydrocodone N-Oxide
CAS 909265-05-0
Analyt- eller komponentnavne Hydrocodone N-Oxide
Forsendelsestilstand Room Temperature
Molekylær formel C18 H21 N O4
Certificeringer/compliance ISO 9001
Synonym (5α)-4,5-Epoxy-3-methoxy-17-methyl-morphinan-6-one 17-Oxide, Hydrocodone N-Oxide
InChI formel InChI=1S/C18H21NO4/c1-19(21)8-7-18-11-4-5-13(20)17(18)23-16-14(22-2)6-3-10(15(16)18)9-12(11)19/h3,6,11-12,17H,4-5,7-9H2,1-2H3/t11-,12+,17-,18-,19?/m0/s1
SMIL COc1ccc2C[C@@H]3[C@@H]4CCC(=O)[C@@H]5Oc1c2[C@]45CC[N+]3(C)[O-]
IUPAC navn (4R,4aR,7aR,12bS)-9-methoxy-3-methyl-3-oxido-1,2,4,4a,5,6,7a,13-octahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-3-ium-7-one
Molekylvægt (g/mol) 315.36
Mængde 25 mg
Formel vægt 315.1471 g/mol