Få mere at vide
Hydrocinnamic-d9 Acid, CDN
Artikelnummer. 30327850
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327850 1 g -
30320303 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327850 Leverandør CDN Leverandørnr. CDND56661G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Phenylpropanoic Acid D9
CAS 93131-15-8
Molekylær formel C9HD9O2
Mængde 1 g
Synonym Benzene-d5-propanoic-d4 acid (9 CI), 3-Phenylpropanoic acid-d9, 3-(Phenyl-d5)propanoic-2,2,3,3-d4 acid, 3-Phenylpropanoic acid D9, β-Phenylpropionic-d9 acid, β-(Phenyl-d5)propionic-α,β-d4 acid, Hydrocinnamic-d9 acid, Hydrocinnamic acid-d9, Benzenepropanoic
InChI formel InChI=1 S/C9H10O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-5 H,6-7H2,(H,10,11)/i1D,2 D,3 D,4 D,5 D,6D2,7D2
SMIL OC(C([2 H])([2 H])C([2 H])([2 H])C1=C([2 H])C([2 H])=C([2 H])C([2 H])=C1[2 H])=O
IUPAC navn 3-(phenyl-d5)propanoic-2,2,3,3-d4 acid
Molekylvægt (g/mol) 159.23
Formel vægt 159.12 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity