missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT™, J.T. Baker™

Catalog Number p-7220830
Click to view available options
Quantity:
50 mL
150 mL
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
Cl
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
Cl

For Laboratory, Research, or Manufacturing Use

  • Management of Change (MOC) category = N
  • (1/10 equiv. = 3.646 g HCl)
  • Makes 0.1 N solution after dilution to 1000 mL
Concentration 0.1 mol/L
Density 1 g/cm³ (20°C)
Packaging DILUT-IT™ Ampule (LDPE)
Normality Passes Test
Quantity 50 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'