Learn More
Hydrochloric Acid, 0.1M, Chem-Lab
For laboratory use, Titrations
Brand: Chem Lab CL0503079010
Additional Details : CAS Number : 7647-01-0 Weight : 10.00000kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Packaging | PE |
---|---|
Quantity | 10L |
Chemical Identifiers
7647-01-0 | |
36.458 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid | |
CHEBI:17883 | |
Cl |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
chlorane |
Specifications
Hydrochloric acid | |
7647-01-0 | |
UN1789 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
3.646g HCl / 1L H2O | |
1.00g/mL | |
313 | |
36.46g/mol | |
0.1N (±0.0002 at 20°C) |
10L | |
ClH | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid | |
Cl | |
chlorane | |
36.458 | |
CHEBI:17883 | |
PE |
Safety and Handling
- Hydrochloric acid 0.1 mol/l
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : HCl/H2O