missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.1M, Chem-Lab
Click to view available options
Quantity:
10 L
20 L
25 L
Packaging:
EP
PE
Specifications
Concentration | 3.646g HCl / 1L H2O |
Density | 1.00g/mL |
Packaging | PE |
Normality | 0.1N (±0.0002 at 20°C) |
Quantity | 25 L |
UN Number | UN1789 |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid |
IUPAC Name | hydrogen chloride |
Formula Weight | 36.46g/mol |
Chemical Name or Material | Hydrochloric acid |
Safety and Handling
Product Identifier
- Hydrochloric acid 0.1 mol/l
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contains : HCl/H2O
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'