Learn More
Hydrobromic Acid Solution, purum p.a., ≥62% (T), Honeywell™
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass Bottle
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Color | Brown-Yellow |
Melting Point | -11°C |
Boiling Point | 126°C |
Density | 1.73g/cu.cm |
Packaging | Glass Bottle |
Quantity | 250 mL |
Physical Form | Clear liquid |
UN Number | UN1788 |
Beilstein | 3587158 |
IUPAC Name | bromane |