Learn More
Hydrazinium Hydroxide, Sigma-Aldrich™
Product Code. 15896380
Change view
Click to view available options
Quantity:
250 mL
1 L
55 kg
Unit Size:
1 L
250mL
55kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896380 1 L 1 L
15869150 250 mL 250mL
15879150 55 kg 55kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15896380 Supplier Sigma Aldrich Supplier No. 8046081000
 Restricted Item
Forventet forsendelse: 26-11-2026
About 100% N2H5OH for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7803-57-8
Specifications
Boiling Point 120.5°C/1013 hPa
Melting Point -51.7°C
Quantity 1 L
Density 1.03 g/cm3 at 20°C
MDL Number MFCD00149931
Synonym Hydrazinium hydroxide, Hydrazine hydrate, Hydrazinium hydroxide
Molecular Weight (g/mol) 32.05 (anhydrous basis)
Grade Research
Chemical Name or Material Hydrazinium Hydroxide