Få mere at vide
Hydantoin-5-13C,1-15N, TRC
Artikelnummer. 30006391
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006391 1 mg
30008641 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006391 Leverandør Toronto Research Chemicals Leverandørnr. TRCH6750031MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189697-61-7
Molekylær formel C213CH4N15NO2
Molekylvægt (g/mol) 102.06
SMIL O=C1[13CH2][15NH]C(=O)N1
Tekniske data
Kemisk navn eller materiale Hydantoin-5-13C,1-15N
Mængde 1 mg
InChI formel InChI=1S/C3H4N2O2/c6-2-1-4-3(7)5-2/h1H2,(H2,4,5,6,7)/i1+1,4+1
Formel vægt 102.0277