Tekniske data
| Kemisk navn eller materiale | Hydantoic Acid |
| CAS | 462-60-2 |
| Molekylær formel | C3H6N2O3 |
| Mængde | 10 g |
| Synonym | hydantoic acid, n-carbamoylglycine, carbamoylglycine, glycoluric acid, n-carbamylglycine, glycine, n-aminocarbonyl, acetic acid, ureido, urea, carboxymethyl, 2-ureidoacetic acid, carbomoylglycine |
| InChI nøgle | KZVRXPPUJQRGFN-UHFFFAOYSA-N |
| SMIL | C(C(=O)O)NC(=O)N |
| IUPAC navn | 2-(carbamoylamino)acetic acid |
| Molekylvægt (g/mol) | 118.092 |
| PubChem CID | 10020 |
| Vis mere |