Learn More
HyClone™ Water, Molecular Biology Grade, Cytiva
Product Code. 10452915
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
6 x 500 mL
6 x 1 L
Unit Size:
1000mL
100mL
500mL
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10452915 6 x 1 L Pack of 6
10275262 100 mL 100mL
10787944 500 mL 500mL
10307052 1000 mL 1000mL
10247783 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10452915 Supplier HyClone products Cytiva Supplier No. SH30538.LS
  • ONLINE EXCLUSIVE PRICE
    1270.00 DKK
    738.27 DKK / Pakke med 6
    Spare 531.73 DKK
    41% Slukket
 Restricted Item
På lager
For critical applications involving nucleotides such as RNA and DNA. HyClone™ Water, Molecular Biology Grade contains no DEPC or other carcinogenic chemicals.
  • Format: Liquid
TRUSTED_SUSTAINABILITY
Specifications
Quantity 6 x 1 L