Learn More
Cytiva HyClone™ Dulbecco's Modified Eagles Medium
Product Code. 10524684
Change view
Click to view available options
Quantity:
500 mL
1000 mL
6 x 500 mL
6 x 1000 mL
With Additives:
Glucose
Glucose, Sodium Pyruvate
Glutamine, Glucose
Glutamine, Ribonucleosides, Deoxyribonucleosides
Glutamine, Sodium Pyruvate
Without Additives:
No Glutamine
No Glutamine, No Phenol Red
No Glutamine, No Phenol Red, No Sodium Pyruvate
No Glutamine, No Sodium Pyruvate
No HEPES
No Phenol Red, No Sodium Pyruvate
None
Unit Size:
1000mL
500mL
Pack of 6
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity With Additives Without Additives Unit Size
10524684 6 x 500 mL Glutamine, Sodium Pyruvate No HEPES Pack of 6
10673313 500 mL Glutamine, Ribonucleosides, Deoxyribonucleosides No Phenol Red, No Sodium Pyruvate 500mL
10402952 1000 mL Glucose No Glutamine, No Sodium Pyruvate 1000mL
11988075 6 x 500 mL Glucose No Glutamine, No Sodium Pyruvate Pack of 6
10544144 6 x 1000 mL Glucose No Glutamine, No Sodium Pyruvate Pack of 6
11511871 1000 mL Glutamine, Sodium Pyruvate None 1000mL
10196223 6 x 1000 mL Glutamine, Sodium Pyruvate None Pack of 6
10443142 500 mL Glucose, Sodium Pyruvate No Glutamine 500mL
10346712 1000 mL Glucose, Sodium Pyruvate No Glutamine 1000mL
10004873 6 x 1000 mL Glucose, Sodium Pyruvate No Glutamine Pack of 6
10359583 6 x 500 mL Glucose, Sodium Pyruvate No Glutamine Pack of 6
10523344 500 mL Glucose, Sodium Pyruvate No Glutamine, No Phenol Red 500mL
11551841 500 mL Glucose No Glutamine, No Sodium Pyruvate 500mL
10658364 1000 mL Glucose No Glutamine, No Phenol Red, No Sodium Pyruvate 1000mL
10749864 500 mL Glutamine, Sodium Pyruvate No HEPES 500mL
10294972 1000 mL Glutamine, Glucose No Phenol Red, No Sodium Pyruvate 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 10524684 Supplier Cytiva Supplier No. SH30243.FS
  • ONLINE EXCLUSIVE PRICE
    1095.00 DKK
    244.62 DKK / Pakke med 6
    Spare 850.38 DKK
    77% Slukket
View all promotions
 Restricted Item
På lager
Designed to support cell culture growth and biomanufacturing processes.
  • Format: Liquid
TRUSTED_SUSTAINABILITY
Specifications
Classification None
Form Liquid
Product Type DMEM/High glucose with L-glutamine, sodium pyruvate
Sterility Sterile
Sterilization Method 0.1 μm Sterile-filtered
With Additives Glutamine, Sodium Pyruvate
Without Additives No HEPES
Quantity 6 x 500 mL
Certifications/Compliance ISO 9001:2000
Content And Storage 2° to 8°C, away from light