Få mere at vide
Cytiva Hyclone™ Donorserum til heste
Mængde:
1 l
Forarbejdning:
Bestrålet
Ingen
Pakningsstørrelse:
Hver
Beskrivelse
Serum Collection:
- From collection to final packaging
- Every step is documented
- Full traceability compliant with current Good Manufacturing Practices (cGMP)
- Extensive quality, performance, and analytical testing
- Aseptically packaged in a Class 100 clean room condition
- Each lot of serum is tested to ensure quality and sterility
- Pooled after filtration and before final packaging
- Consistent quality from the first bottle filled to the last
- Increases consistency and minimizes variability with a lot of serum
Tekniske data
Tekniske data
| Yderligere analyser | Albumin |
| Oprindelsesland | United States |
| Form | Liquid |
| Produkttype | Equine Serum |
| Mængde | 1 l |
| Sterilitet | Sterile |
| Steriliseringsmetode | Sequential Triple-filtered, 100 nm |
| Forarbejdning | Bestrålet |
| Testet for | BVDV, Rabies Virus, Reovirus, Cytopathogenic Agents |
| Varme inaktiveret | No |
| Vis mere |