Learn More
Cytiva HyClone™ Cell Boost™ 4 Supplement
Product Code. 16685795
Change view
Click to view available options
Reconstituted Volume:
500 mL
1000 mL
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Reconstituted Volume Unit Size
16685795 1000 mL 1 L
16534581 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16685795 Supplier Cytiva Supplier No. SH31116.02
  • ONLINE EXCLUSIVE PRICE
    1770.00 DKK
    1190.00 DKK / 1 L
    Spare 580.00 DKK
    32% Slukket
 Restricted Item
Forventet forsendelse: 25-11-2026
Fed-batch media supplement containing amino acids, vitamins, glucose, trace elements, and growth factors for CHO cells
  • Chemically Defined (CD), Animal-Derived Component-Free (ADCF), Protein-Free, and Hydrolysate-Free
  • Supports fed-batch cultures with amino acids, vitamins, glucose, trace elements, and growth factors
  • Developed for recombinant protein production with various cell lines including Chinese Hamster Ovary (CHO) cell line
  • Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
  • Manufactured to meet cGMP manufacturing standards and QC specifications
  • Available as a ready-to-use liquid feed solution
TRUSTED_SUSTAINABILITY
Specifications
Cell Line CHO
Form Liquid
Product Type Cell Boost Supplement
Format Bottle
For Use With (Application) Improving Productivity and Cell Yield in Fed-Batch Cultures
Product Line HyClone
Reconstituted Volume 1000 mL