Learn More
Cytiva HyClone™ AdvanceSTEM™ Mesenchymal Stem Cell Expansion Kit
Product Code. 12300133
Change view
Click to view available options
Quantity:
1000 mL
500 mL
Kit - to make 11 mL complete media
Unit Size:
1 set
1000mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12300133 Kit - to make 11 mL complete media 1 set
11576351 1000 mL 1000mL
11566351 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12300133 Supplier Cytiva Supplier No. SH30878.01
  • ONLINE EXCLUSIVE PRICE
    982.00 DKK
    734.38 DKK / 1 sæt
    Spare 247.62 DKK
    25% Slukket
View all promotions
 Restricted Item
På lager
Support expansion and maintenance of undifferentiated human mesenchymal stem cells (hMSCs) with HyClone™ AdvanceSTEM™ Mesenchymal Stem Cell Expansion Kit.
TRUSTED_SUSTAINABILITY
Specifications
Cell Type Mesenchymal Stem Cells
Classification None
Form Liquid
Product Type Stem Cell Medium Supplement
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Quantity Kit - to make 11 mL complete media
Content And Storage SH3878.1, SH3879.2
Description Mesenchymal Stem Cell Growth Supplement
Culture Type Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)