missing translation for 'onlineSavingsMsg'
Få mere at vide

Hycanthone, MedChemExpress

Artikelnummer. 16418668
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16418668 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16418668 Leverandør Medchem Express Leverandørnr. HYB1099

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Hycanthone is a thioxanthenone DNA intercalator and inhibits RNA synthesis as well as the DNA topoisomerases I and II. Hycanthone inhibits nucleic acid biosynthesis and inhibits apurinic endonuclease-1 (APE1) by direct protein binding with a KD of 10 nM. Hycanthone is a bioactive metabolite of Lucanthone (HY-B2098) and has anti-schistosomal agent.

Hycanthone is a thioxanthenone DNA intercalator and inhibits RNA synthesis as well as the DNA topoisomerases I and II. Hycanthone inhibits nucleic acid biosynthesis and inhibits apurinic endonuclease-1 (APE1) by direct protein binding with a KD of 10 nM. Hycanthone is a bioactive metabolite of Lucanthone (HY-B2098) and has anti-schistosomal agent.

Tekniske data

Kemisk navn eller materiale Hycanthone
Fysisk form Solid
CAS 3105-97-3
Farve Deep Yellow
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C20H24N2O2S
Til brug med (applikation) COVID-19-anti-virus
Opløselighedsinformation DMSO : 12.5 mg/mL (35.07 mM; Need ultrasonic)
SMIL O=C1C2=C(SC3=C1C=CC=C3)C(CO)=CC=C2NCCN(CC)CC
Molekylvægt (g/mol) 356.48
Formel vægt 356.48
Procent renhed 98.85%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.