Learn More
Hyaluronic acid sodium salt, BLD Pharm™
Product Code. 31328191
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31328191 5 g
31326610 100 g
31326995 25 g
31326072 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31328191 Supplier BLD Pharm Supplier No. BD1340655g
 Restricted Item
Forventet forsendelse: 25-11-2026
Mol wt 750,000 to 1,000,000
Specifications
CAS 9067-32-7
MDL Number MFCD00875848
Quantity 5 g
SMILES NONE
Chemical Name or Material Hyaluronic acid sodium salt