Learn More
Hamilton™ HVP Panel Mount Valves
Product Code. 11532465
Change view
Click to view available options
Material:
Aluminum (Housing)
Unit Size:
Each
Product Code. Material Unit Size
11532465 Aluminum (Housing) Each
1 options
Product Code. 11532465 Supplier Hamilton™ Supplier No. 86779
  • ONLINE EXCLUSIVE PRICE
    1135.00 DKK
    991.25 DKK / Pakke med 1
    Spare 143.75 DKK
    12% Slukket
Forventet forsendelse: 10-12-2026
Can be mounted into panels allowing for custom installation into control panels or instruments.
  • Manual actuated
  • Basic unit for control of liquids
  • Ideal for low pressure applications
Specifications
Volume (Metric) Sample 14 μL
Description 4-4 Model, 4 Ports, Loop Flow Path
Material Aluminum (Housing)
Temperature (Metric) Maximum 38°C