Learn More
Hamilton™ HV 4-2 Valve, 4 ports, 90 deg Flow Path, aluminum housing
Product Code. 11570052
Change view
Click to view available options
Manual valve
Product Code.
11570052 Manual valve
1 options
Product Code. 11570052 Supplier Hamilton™ Supplier No. 86731
  • ONLINE EXCLUSIVE PRICE
    1270.00 DKK
    1143.24 DKK / Pakke med 1
    Spare 126.76 DKK
    9% Slukket
Forventet forsendelse: 11-12-2026
Providing an economical and flexible method of custom-building a fluid flow control system.
  • Chemically inert flow path
  • Ideally suited for low pressure applications
  • Leak-tight seals