missing translation for 'onlineSavingsMsg'
Få mere at vide

Huperzine C, TRC

Artikelnummer. 30404502
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30404502 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30404502 Leverandør Toronto Research Chemicals Leverandørnr. TRCH6739201MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Huperzine C
CAS 163089-71-2
Molekylær formel C15 H18 N2 O
Mængde 1 mg
Synonym (5R,9R,11R)- 5-Amino-11-ethenyl-5,6,9,10-tetrahydro-7-methyl-5,9-methanocycloocta[b]pyridin-2(1H)-one
InChI formel InChI=1S/C15H18N2O/c1-3-11-10-6-9(2)8-15(11,16)12-4-5-14(18)17-13(12)7-10/h3-6,10-11H,1,7-8,16H2,2H3,(H,17,18)
SMIL CC1=CC2CC3=C(C=CC(=O)N3)C(N)(C1)C2C=C
Molekylvægt (g/mol) 242.316
Formel vægt 242.142
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.