missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Proteintech™ HumanKine™ Recombinant Human VEGF121 Protein
Shop alle Proteintech produkter
Klik for at se tilgængelige muligheder
Mængde:
10 μg
100 μg
1000 μg
Beskrivelse
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Tekniske data
Tekniske data
| Gen-id (Entrez) | 7422 |
| Molekylvægt (g/mol) | 37 kDa, homodimer; 50 kDa, homotrimer, glycosylated |
| Mængde | 10 μg |
| Arter | Human |
| Rekonstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
| Indhold og opbevaring | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
| Aktivitet | Typically ≤ 15 ng/mL EC50 |
| Udtrykssystem | Human embryonic kidney 293 (HEK293) human cell expression system |
| Form | Lyophilized |
| Renhed eller kvalitetsklasse | > 95% |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion