Få mere at vide
Proteintech™ HumanKine™ Recombinant Human Sonic Hedgehog (SHH) Protein
Shop alle Proteintech produkter
Mængde:
10 μg
100 μg
5 μg
Pakningsstørrelse:
100µg
5µg
Beskrivelse
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Tekniske data
Tekniske data
| Gen-id (Entrez) | 6469 |
| Molekylvægt (g/mol) | 20 to 24 kDa |
| Mængde | 100 μg |
| Arter | Human |
| Rekonstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile water containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
| Indhold og opbevaring | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
| Aktivitet | Typically ≤ 350 ng/mL EC50 |
| Udtrykssystem | Human embryonic kidney 293 (HEK293) human cell expression system |
| Form | Lyophilized |
| Renhed eller kvalitetsklasse | > 90% |