Learn More
Abnova™ Human ZNF398 Partial ORF (NP_065832.1, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057541-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the Kruppel family of C2H2-type zinc-finger transcription factor proteins. The encoded protein acts as a transcriptional activator. Two transcript variants encoding distinct isoforms have been identified for this gene. Other transcript variants have been described, but their full length sequence has not been determined. [provided by RefSeq]
Sequence: MKGNYESLISMDYAINQPDVLSQIQPEGEHNTEDQAGPEESEIPTDPSEEPGISTSDILSWIKQEEEPQVGAPPESKESDVYKSTYADEESpecifications
NP_065832.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKGNYESLISMDYAINQPDVLSQIQPEGEHNTEDQAGPEESEIPTDPSEEPGISTSDILSWIKQEEEPQVGAPPESKESDVYKSTYADEE | |
RUO | |
ZNF398 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
57541 | |
ZNF398 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1339/P51/P71/ZER6 | |
ZNF398 | |
Recombinant | |
wheat germ expression system |