missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human ZNF222 Control Fragment Recombinant Protein

Artikelnummer. 30181458
Click to view available options
Mængde:
100 μL
This item is not returnable. View return policy

Product Code. 30181458

Brand: Invitrogen™ RP99763

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (38%), Rat (38%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52047 (PA5-52047. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TRUSTED_SUSTAINABILITY

Specifications

Adgangsnummer Q9UK12
Koncentration ≥5.0 mg/mL
Til brug med (applikation) Blocking Assay, Control
Formulering 1 M urea, PBS with no preservative; pH 7.4
Gen-id (Entrez) 7673
Navn Human ZNF222 Control Fragment
Mængde 100 μL
Regulatorisk status RUO
Gene Alias Zinc finger protein 222; ZNF222
Fælles navn ZNF222
Gen symbol ZNF222
Konjugeret Unconjugated
Arter Human
Rekombinant Recombinant
Protein tag His-ABP-tag
Sekvens AQRKLYRDVMLENFRNLLSVGGKIQTEMETFPEAGTHEEFSCKQIWEQIASDLTRSQDTTISNSQLFEQDDNPSQIKARLSTVHTREKPFQGENCKQFFSDVSFFDLPQ
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Udtrykssystem E. coli
Form Liquid
Renhed eller kvalitetsklasse >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.