Få mere at vide
Invitrogen™ Human ZNF211 (aa 175-248) Control Fragment Recombinant Protein
Artikelnummer. 30205763
Skift visning
Mængde:
100 μL
Artikelnummer. Mængde
30205763 100 μL
1 options
Artikelnummer. 30205763 Leverandør Invitrogen™ Leverandørnr. RP106254
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (33%), Rat (33%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65591 (PA5-65591. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY
Tekniske data
Adgangsnummer Q13398
Koncentration ≥5.0 mg/mL
Til brug med (applikation) Blocking Assay, Control
Formulering 1 M urea, PBS with no preservative; pH 7.4
Gen-id (Entrez) 10520
Navn Human ZNF211 (aa 175-248) Control Fragment
Mængde 100 μL
Regulatorisk status RUO
Gene Alias C2H2-25; CH2H2-25; Zinc finger protein 211; zinc finger protein C2H2-25; ZNF211; ZNF-25; ZNFC25
Fælles navn ZNF211
Gen symbol ZNF211
Konjugeret Unconjugated
Arter Human
Rekombinant Recombinant
Protein tag His-ABP-tag
Sekvens SCIVHVSEKPFTCREIRKDFLANMRFLHQDATQTGEKPNNSNKCAVAFYSGKSHHNWGKCSKAFSHIDTLVQDQ
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Udtrykssystem E. coli
Form Liquid
Renhed eller kvalitetsklasse >80% by SDS-PAGE and Coomassie blue staining