Learn More
Abnova™ Human ZDHHC15 Full-length ORF (NP_659406.1, 1 a.a. - 337 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00158866-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the DHHC palmitoyltransferase family. Mutations in this gene are associated with mental retardatio X-linked type 91 (MRX91). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MRRGWKMALSGGLRCCRRVLSWVPVLVIVLVVLWSYYAYVFELCLVTVLSPAEKVIYLILYHAIFVFFTWTYWKSIFTLPQQPNQKFHLSYTDKERYENEERPEVQKQMLVDMAKKLPVYTRTGSGAVRFCDRCHLIKPDRCHHCSVCAMCVLKMDHHCPWVNNCIGFSNYKFFLQFLAYSVLYCLYIATTVFSYFIKYWRGELPSVRSKFHVLFLLFVACMFFVSLVILFGYHCWLVSRNKTTLEAFCTPVFTSGPEKNGFNLGFIKNIQQVFGDKKKFWLIPIGSSPGDGHSFPMRSMNESQNPLLANEETWEDNEDDNQDYPEGSSSLAVETETSpecifications
NP_659406.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ31812/MGC119974/MGC119975/MGC119976/MRX91 | |
ZDHHC15 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
158866 | |
ZDHHC15 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRRGWKMALSGGLRCCRRVLSWVPVLVIVLVVLWSYYAYVFELCLVTVLSPAEKVIYLILYHAIFVFFTWTYWKSIFTLPQQPNQKFHLSYTDKERYENEERPEVQKQMLVDMAKKLPVYTRTGSGAVRFCDRCHLIKPDRCHHCSVCAMCVLKMDHHCPWVNNCIGFSNYKFFLQFLAYSVLYCLYIATTVFSYFIKYWRGELPSVRSKFHVLFLLFVACMFFVSLVILFGYHCWLVSRNKTTLEAFCTPVFTSGPEKNGFNLGFIKNIQQVFGDKKKFWLIPIGSSPGDGHSFPMRSMNESQNPLLANEETWEDNEDDNQDYPEGSSSLAVETET | |
RUO | |
ZDHHC15 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |