missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZDHHC15 Full-length ORF (NP_659406.1, 1 a.a. - 337 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00158866-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16178873

  • 2665.00 DKK / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene belongs to the DHHC palmitoyltransferase family. Mutations in this gene are associated with mental retardatio X-linked type 91 (MRX91). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MRRGWKMALSGGLRCCRRVLSWVPVLVIVLVVLWSYYAYVFELCLVTVLSPAEKVIYLILYHAIFVFFTWTYWKSIFTLPQQPNQKFHLSYTDKERYENEERPEVQKQMLVDMAKKLPVYTRTGSGAVRFCDRCHLIKPDRCHHCSVCAMCVLKMDHHCPWVNNCIGFSNYKFFLQFLAYSVLYCLYIATTVFSYFIKYWRGELPSVRSKFHVLFLLFVACMFFVSLVILFGYHCWLVSRNKTTLEAFCTPVFTSGPEKNGFNLGFIKNIQQVFGDKKKFWLIPIGSSPGDGHSFPMRSMNESQNPLLANEETWEDNEDDNQDYPEGSSSLAVETET
Specifications

Specifications

NP_659406.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ31812/MGC119974/MGC119975/MGC119976/MRX91
ZDHHC15
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
158866
ZDHHC15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRRGWKMALSGGLRCCRRVLSWVPVLVIVLVVLWSYYAYVFELCLVTVLSPAEKVIYLILYHAIFVFFTWTYWKSIFTLPQQPNQKFHLSYTDKERYENEERPEVQKQMLVDMAKKLPVYTRTGSGAVRFCDRCHLIKPDRCHHCSVCAMCVLKMDHHCPWVNNCIGFSNYKFFLQFLAYSVLYCLYIATTVFSYFIKYWRGELPSVRSKFHVLFLLFVACMFFVSLVILFGYHCWLVSRNKTTLEAFCTPVFTSGPEKNGFNLGFIKNIQQVFGDKKKFWLIPIGSSPGDGHSFPMRSMNESQNPLLANEETWEDNEDDNQDYPEGSSSLAVETET
RUO
ZDHHC15
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers