Learn More
Abcam Human ZC3HAV1 (Zinc finger antiviral protein) knockout A549 cell line
Product Code. 32142405 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
32142405 2 x 1,000,000 Cells Pack of 2
1 options
Product Code. 32142405 Supplier Abcam Supplier No. ab2669522x1000000Cells_
 Restricted Item
Forventet forsendelse: 30-11-2026
ZC3HAV1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp insertion in exon 4. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells: selec...
ZC3HAV1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp insertion in exon 4. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells
Regulatory Status RUO