Learn More
Abnova™ Human WDR57 Partial ORF (NP_004805.1, 262 a.a. - 357 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009410-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a component of the U5 small nuclear ribonucleoprotein (snRNP) particle. The U5 snRNP is part of the spliceosome, a multiprotein complex that catalyzes the removal of introns from pre-messenger RNAs. [provided by RefSeq]
Sequence: WDVRPFAPKERCVKIFQGNVHNFEKNLLRCSWSPDGSKIAAGSADRFVYVWDTTSRRILYKLPGHAGSINEVAFHPDEPIIISASSDKRLYMGEIQSpecifications
NP_004805.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
WDVRPFAPKERCVKIFQGNVHNFEKNLLRCSWSPDGSKIAAGSADRFVYVWDTTSRRILYKLPGHAGSINEVAFHPDEPIIISASSDKRLYMGEIQ | |
RUO | |
SNRNP40 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9410 | |
WDR57 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
40K/FLJ41108/HPRP8BP/MGC1910/PRP8BP/PRPF8BP/RP11-490K7.3/SPF38/WDR57 | |
SNRNP40 | |
Recombinant | |
wheat germ expression system |