missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human WDR36 Partial ORF (NP_644810, 853 a.a. - 951 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_644810 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 134430 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16141857
|
Abnova™
H00134430-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16131857
|
Abnova™
H00134430-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Mutations in this gene have been associated with adult-onset primary open-angle glaucoma (POAG). [provided by RefSeq]
Sequence: SGIETELRSLSPDCGGSIEVMQSFLKMIGMMLDRKRDFELAQAYLALFLKLHLKMLPSEPVLLEEITNLSSQVEENWTHLQSLFNQSMCILNYLKSALLSpecifications
NP_644810 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686I1650/GLC1G/TA-WDRP/TAWDRP/UTP21 | |
WDR36 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
134430 | |
WDR36 (Human) Recombinant Protein (Q01) | |
SGIETELRSLSPDCGGSIEVMQSFLKMIGMMLDRKRDFELAQAYLALFLKLHLKMLPSEPVLLEEITNLSSQVEENWTHLQSLFNQSMCILNYLKSALL | |
RUO | |
WDR36 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |