missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human WBSCR27 Full-length ORF (AAH30295.1, 1 a.a. - 245 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00155368-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16178813

  • 4040.00 DKK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein belonging to ubiE/COQ5 methyltransferase family. The gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.22-q11.23. [provided by RefSeq]

Sequence: MAQEEGGSLPEVRARVRAAHGIPDLAQKLHFYDRWAPDYDQDVATLLYRAPRLAVDCLTQALPGPPHSALILDVACGTGLVAAELRAPGFLQLHGVDGSPGMLEQARAPGLYQRLSLCTLGQEPLPSPEGTFDAVLIVGALSDGQVPCNAIPELHVTKPGGLVCLTTRTNWSNLQYKEALEATLDRLEQAGMWEGLVAWPVDRLWTAGSWLPPSWWWYPASLPRMASSPALSTCTESGRRPRLRK
Specifications

Specifications

AAH30295.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC40131
WBSCR27
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
155368
WBSCR27 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAQEEGGSLPEVRARVRAAHGIPDLAQKLHFYDRWAPDYDQDVATLLYRAPRLAVDCLTQALPGPPHSALILDVACGTGLVAAELRAPGFLQLHGVDGSPGMLEQARAPGLYQRLSLCTLGQEPLPSPEGTFDAVLIVGALSDGQVPCNAIPELHVTKPGGLVCLTTRTNWSNLQYKEALEATLDRLEQAGMWEGLVAWPVDRLWTAGSWLPPSWWWYPASLPRMASSPALSTCTESGRRPRLRK
RUO
WBSCR27
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers