missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ Human Vitronectin, Animal-Free Recombinant Protein, PeproTech®

Artikelnummer. 17873723
Klik for at se tilgængelige muligheder
Mængde:
1 mg
100 μg
500 μg
Pakningsstørrelse:
1mg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 17873723

Brand: Gibco™ AF14009500UG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Recombinant Human Vitronectin is a 459 amino acid, single-chain, monomeric protein, which migrates at an apparent molecular weight of 75 kDa by SDS-PAGE under reducing conditions. The calculated molecular weight of Recombinant Human Vitronectin is 52.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

VTN is a member of the pexin family. This protein is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. The protein is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P04004
Til brug med (applikation) Functional Assay
Formulering protein with no preservative
Gen-id (Entrez) 7448
Molekylvægt (g/mol) 52.2 kDa
Navn Human Vitronectin, Animal-Free
Mængde 500 μg
Kilde HEK293 cells
Regulatorisk status RUO
Endotoksin koncentration <0.1 EU/ μg
Gene Alias Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN
Fælles navn Vitronectin
Gen symbol VTN
Biologisk aktivitet Recombinant human Vitronectin promotes attachment of hESC and iPSC in serum-free, feeder conditions at 5 ug/ml
Konjugeret Unconjugated
Rekombinant Recombinant
Sekvens DQESCKGRCT EGFNVDKKCQ CDELCSYYQS CCTDYTAECK PQVTRGDVFT MPEDEYTVYD DGEEKNNATV HEQVGGPSLT SDLQAQSKGN PEQTPVLKPE EEAPAPEVGA SKPEGIDSRP ETLHPGRPQP PAEEELCSGK PFDAFTDLKN GSLFAFRGQY CYELDEKAVR PGYPKLIRDV WGIEGPIDAA FTRINCQGKT YLFKGSQYWR FEDGVLDPDY PRNISDGFDG IPDNVDAALA LPAHSYSGRE RVYFFKGKQY WEYQFQHQPS QEECEGSSLS AVFEHFAMMQ RDSWEDIFEL LFWGRTSAGT RQPQFISRDW HGVPGQVDAA MAGRIYISGM APRPSLAKKQ RFRHRNRKGY RSQRGHSRGR NQNSRRPSRA TWLSLFSSEE SNLGANNYDD YRMDWLVPAT CEPIQSVFFF SGDKYYRVNL RTRRVDTVDP PYPRSIAQYW LGCPAPGHL
Indhold og opbevaring -20°C
Udtrykssystem HEK293 cells
Form Lyophilized
Renhed eller kvalitetsklasse ≥ 95% by SDS-PAGE gel and HPLC analyses.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.